Learn More
anti-CCR6, PE, Clone: 876515, R&D Systems™
Manufacturer:  R&D Systems FAB8320P100
 View more versions of this product
Catalog No. FAB8320P100
Add to cart
Specifications
Specifications
CCR6
876515
Flow Cytometry 10 uL/10^6 cells
NP_001013163
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22
Mouse
IgG2b
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Stains rat CCR6 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR6
NS0 mouse myeloma cell line transfected with rat CCR6, Met1-Met366, Accession # NP_001013163
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1235
Rat