Learn More
anti-CCR6, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131236
Catalog No. NBP131236
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CCR6
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
CCR6
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CCR6. The exact sequence is proprietary.
42 kDa
100 ul
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary
1235
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Predicted cross-reactivity based on sequence identity: Rhesus Monkey (86%). Not yet tested in any other species.