Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR7, Alexa Fluor 700, Clone: 150503, R&D Systems™

Manufacturer:  R&D Systems FAB197N025

 View more versions of this product

Catalog No. FAB197N025


Add to cart

Description

Description

Host Species: Mouse
Clone: 150503
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: Human CCR7 transfectants. Met1-Pro378

Applications:

Flow Cytometry

Specifications

Specifications

CCR7
150503
Flow Cytometry 5 uL/10^6 cells
AAA58615
BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
Mouse
IgG2a
25 Tests
Primary
1236
Human
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR7
Human CCR7 transfectants Met1-Pro378 Accession # AAA58615
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CCR7.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit