Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR7, Fluorescein, Clone: 150503, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  R&D Systems FAB197F100

 View more versions of this product

Catalog No. FAB197F100


Add to cart

Description

Description

Host Species: Mouse
Clone: 150503
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: Human CCR7 transfectants. Met1-Pro378

Applications:

Flow Cytometry

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

CCR7
150503
Flow Cytometry 10 uL/10^6 cells
AAA58615
BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
Mouse
IgG2a
100 Tests
Primary
1236
Human
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR7
Human CCR7 transfectants, Met1-Pro378, Accession # AAA58615
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human CCR7.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit