Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR7, PerCP, Clone: 150503, R&D Systems™

Manufacturer:  R&D Systems FAB197C100

 View more versions of this product

Catalog No. FAB197C100

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Host Species: Mouse
Clone: 150503
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: Human CCR7 transfectants. Met1-Pro378

Applications:

Flow Cytometry

Specifications

Specifications

PerCP
AAA58615
100 Tests
Monoclonal
Supplied in a saline solution containing BSA and Sodium Azide.
Purified
Primary
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit