Learn More
anti-CCR8, APC, Clone: 1055c, R&D Systems™
Manufacturer:  R&D Systems FAB8324A100
 View more versions of this product
Catalog No. FAB8324A100
Add to cart
Specifications
Specifications
CCR8
1055C
Flow Cytometry 10 uL/10^6 cells
NP_031746
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1
Rabbit
IgG
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Stains mouse CCR8 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
CCR8
HEK293 human embryonic kidney cell line transfected with mouse CCR8, Accession # NP_031746
Protein A or G purified from cell culture supernatant
RUO
Primary
1237
Mouse
Documents
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'