Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR8, PE, Clone: 1055c, R&D Systems
Manufacturer: R&D Systems FAB8324P025
Specifications
CCR8 | |
1055C | |
Flow Cytometry 10 uL/10^6 cells | |
NP_031746 | |
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1 | |
Rabbit | |
IgG | |
25 Tests | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
Monoclonal | |
Stains mouse CCR8 transfectants but not irrelevant transfectants in flow cytometry. |
Flow Cytometry | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Affinity Purified | |
CCR8 | |
HEK293 human embryonic kidney cell line transfected with mouse CCR8 Accession # NP_031746 | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
1237 | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title