Learn More
anti-CCR8, PE, Clone: 1055c, R&D Systems™
Manufacturer:  R&D Systems FAB8324P100
 View more versions of this product
Catalog No. FAB8324P100
Add to cart
Specifications
Specifications
CCR8
1055C
Flow Cytometry 10 uL/10^6 cells
NP_031746
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1
Rabbit
IgG
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Stains mouse CCR8 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
CCR8
HEK293 human embryonic kidney cell line transfected with mouse CCR8, Accession # NP_031746
Protein A or G purified from cell culture supernatant
RUO
Primary
1237
Mouse