Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR8, PE, Clone: 191704, R&D Systems
Manufacturer: R&D Systems FAB1429P100
Description
Host Species: Rat
Clone: 191704
Isotype: IgG2B
Species Reactivity: Hu, Rt
Immunogen: BaF3 mouse pro-B cell line transfected with human CCR8. Met1-Leu355
Applications:
Flow Cytometry
Specifications
CCR8 | |
191704 | |
Flow Cytometry 10 uL/10^6 cells | |
P51685 | |
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1 | |
Rat | |
IgG2b | |
100 Tests | |
Primary | |
1237 | |
Human, Rat |
Flow Cytometry | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Purified | |
CCR8 | |
BaF3 mouse pro-B cell line transfected with human CCR8 Met1-Leu355 Accession # P51685 | |
Protein A or G purified from hybridoma culture supernatant | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
Monoclonal | |
Detects human CCR8. Neutralizes the bioactivity of human CCR8 and specifically stains human CCR8-transfected cell lines but not with the parent cell lines. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title