Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR9, APC, Clone: 242503, R&D Systems™

Manufacturer:  R&D Systems FAB2160A025

 View more versions of this product

Catalog No. FAB2160A025


Add to cart

Description

Description

Host Species: Rat
Clone: 242503
Isotype: IgG2B
Species Reactivity: Ms, Rt
Immunogen: Y3 rat myeloid cell line transfected with mouse CCR9. Met1-Leu369

Applications:

Flow Cytometry

Specifications

Specifications

CCR9
242503
Flow Cytometry 10 uL/10^6 cells
Q9WUT7
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Rat
IgG2b
25 Tests
Primary
10803
Mouse, Rat
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR9
Y3 rat myeloid cell line transfected with mouse CCR9, Met1-Leu369, Accession # Q9WUT7
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse and rat CCR9 in flow cytometry. Stains mouse CCR9-transfected cells but not irrelevant transfectants.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit