Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR9, Biotin, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128644

Catalog No. NBP128644


Add to cart

Description

Description

CCR9 Polyclonal antibody specifically detects CCR9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CCR9
1.0mg/mL
Western Blot 2 ug/ml, ELISA 1:100000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 8 ug/ml
Affinity Purified
CCR9
Synthetic peptide: IPGMFDDFSYDSTASTDDYMNLNFSSFF, corresponding to amino acids 10-37 of Mouse CCR9.
Immunogen affinity purified
RUO
Primary
10803
Human, Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Biotin
10 mM KHPO4 and 140 mM NaCl with 0.05% Sodium Azide
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Goat
IgG
0.05 mg
Store at -80C. Avoid freeze-thaw cycles.
Polyclonal
Peptide sequence is < 50 % identical to other mouse chemokine receptors in this region.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only