Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR9, PE, Clone: 112509, R&D Systems™

Manufacturer:  R&D Systems FAB179P100

 View more versions of this product

Catalog No. FAB179P100


Add to cart

Description

Description

Host Species: Mouse
Clone: 112509
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: BaF3 mouse pro-B cell line transfected with human CCR9. Met1-Leu369

Applications:

Flow Cytometry

Specifications

Specifications

CCR9
112509
Flow Cytometry 10 uL/10^6 cells
P51686
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Mouse
IgG2a
100 Tests
Primary
10803
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CCR9
BaF3 mouse pro-B cell line transfected with human CCR9, Met1-Leu369, Accession # P51686
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Specifically stains human CCR9 transfectants and MOLT-4 cells. It does not react with the parental BaF3 cells or human CCR7-transfected BaF3 cells.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit