Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR9, PE, Clone: 112509, R&D Systems
Manufacturer: R&D Systems FAB179P025
Description
Host Species: Mouse
Clone: 112509
Isotype: IgG2A
Species Reactivity: Hu
Immunogen: BaF3 mouse pro-B cell line transfected with human CCR9. Met1-Leu369
Applications:
Flow Cytometry
Specifications
CCR9 | |
112509 | |
Flow Cytometry 10 uL/10^6 cells | |
P51686 | |
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
Mouse | |
IgG2a | |
25 Tests | |
Primary | |
10803 | |
Human |
Flow Cytometry | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Purified | |
CCR9 | |
BaF3 mouse pro-B cell line transfected with human CCR9, Met1-Leu369, Accession # P51686 | |
Protein A or G purified from hybridoma culture supernatant | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
Monoclonal | |
Specifically stains human CCR9 transfectants and MOLT-4 cells. It does not react with the parental BaF3 cells or human CCR7-transfected BaF3 cells. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title