Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CCR9, PerCP, Clone: 112509, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB179C025
This item is not returnable.
View return policy
Description
CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry.Specifications
CCR9 | |
Monoclonal | |
PerCP | |
P51686 | |
CCR9 | |
BaF3 mouse pro-B cell line transfected with human CCR9 Met1-Leu369 Accession # P51686 | |
25 Tests | |
10803 | |
Human | |
Purified |
Flow Cytometry | |
112509 | |
Flow Cytometry 10 uL/10^6 cells | |
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Specifically stains human CCR9 transfectants and MOLT-4 cells. It does not react with the parental BaF3 cells or human CCR7-transfected BaF3 cells. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction