Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRN4L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124489

Catalog No. PIPA124489


Add to cart

Description

Description

PA1-24489 detects CCRN4L in Human samples. PA1-24489 has been successfully used in Immunohistochemistry and Western Blot procedures. PA1-24489 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human CCRN4L. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CCRN4L is highly similar to Nocturnin, a gene identified as a circadian clock regulated gene in Xenopus laevis. This protein and Nocturnin protein share similarity with the C-terminal domain of a yeast transcription factor, carbon catabolite repression 4 (CCR4). The mRNA abundance of a similar gene in mouse has been shown to exhibit circadian rhythmicity, which suggests a role for this protein in clock function or as a circadian clock effector.
Specifications

Specifications

CCRN4L
Polyclonal
Unconjugated
NOCT
Liquid
NOCT
Synthetic peptide region of human CCRN4L.
Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9UK39
CCR4L, MGC142054, MGC142060, MGC4120817, MGC78549, NOC
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
25819
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit