Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

copper chaperone for superoxide dismutase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant CCS.

Manufacturer:  Abnova Corporation H00009973A01

Catalog No. 89-004-459


Add to cart

Description

Description

Copper chaperone for superoxide dismutase specifically delivers Cu to copper/zinc superoxide dismutase and may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor. [provided by RefSeq

Sequence: ADGRAIFRMEDEQLKVWDVIGRSLIIDEGEDDLGRGGHPLSKITGNSGERLACGIIARSAGLFQNPKQICSCDGLTIWEERGRPIAGKGRKESAQPPAHL
Specifications

Specifications

copper chaperone for superoxide dismutase
Unconjugated
50% glycerol
NM_005125
MGC138260
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
9973
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant CCS.
CCS
Serum
CCS
CCS (NP_005116, 175 a.a. ~ 274 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit