Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNYL1 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525634

Catalog No. PIPA525634

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CCNYL1 belongs to the cyclin family, Cyclin Y subfamily and contains 1 cyclin N terminal domain. The specific function of CCNYL1 is not yet known.
Specifications

Specifications

CCNYL1
Polyclonal
PBS with 0.09% sodium azide
Q8N7R7
cyclin-Y-like protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100760997, 151195
Immunohistochemistry, Western Blot
Unconjugated
CCNYL1
Liquid
CCNYL1
KLH conjugated synthetic peptide between 41-70 amino acids from the N-terminal region of human CCYL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit