Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD10/CD19 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Supplier: Thermo Scientific MA112188
Description
This antibody is a monoclonal mixture of FITC-conjugated CD10 (clone RXB.14, isotype IgG1) and PE-conjugated CD19 (clone 1G9, isotype IgG1).
CD10/CD19 Cocktail antibody specifically detects CD10/CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD10/CD19 | |
Cocktail | |
FITC/PE | |
MME | |
atriopeptidase; AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; CALLA; CD10; CD19; CD19 molecule; CMT2T; Common acute lymphocytic leukemia antigen; CVID3; differentiation antigen CD19; Enkephalinase; EPN; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; Neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase; T-cell surface antigen Leu-12 | |
Mouse | |
Protein A/G | |
RUO | |
4311, 930 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
RXB.14, 1G9, 1G9 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P08473, P15391 | |
CD19, MME | |
White blood cells. | |
50 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction