Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti CD10, Clone: J5, Beckman Coulter™

$391.68 - $735.42

Specifications

Antigen CD10
Clone J5
Format Affinity Purified
Host Species Murine
Isotype IgG2a
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CO6602463
View Documents
Coulter Corporation
6602463
100 Tests Each for $735.42
Add to cart
 
CO6604107
View Documents
Coulter Corporation
6604107
50 Tests Each for $391.68
Add to cart
 
Description

Description

The CD10 antigen is refered to as the Common Acute Lymphoblastic Leukemia Antigen (CALLA). It is a type II integral membrane protein of 100 kDa, identified as the human membrane-associated neutral endopeptidase (EC3.4.24.11). It is expressed on uncommitted lymphoid precursors. CD10 expression is lost as cells enter the T lineage. In the B lineage, CD10 expression is lost later in ontogeny, as cells acquire surface Ig expression. It is also expressed on activated and proliferating B cells in the germinal center, and on neutrophils as well as on bone marrow stromal cells. It is also expressed on a number of other cells of epithelial origin.

Host Species: Mouse
Clone: J5
Isotype: IgG2a
Cross Reactivity: Human

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

CD10
Affinity Purified
IgG2a
Primary
J5
Murine
Monoclonal
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit