Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD101 Monoclonal Antibody (307707), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA524021
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
CD101 Monoclonal antibody specifically detects CD101 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD101 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
A8E0Y8 | |
CD101 | |
CHO-derived recombinant mouse IGSF2/CD101 Gln21-Tyr1033 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
307707 | |
Unconjugated | |
CD101 | |
Cd101; CD101 antigen; CD101 molecule; cell surface glycoprotein V7; EWI immunoglobulin subfamily member EWI-101; EWI101; EWI-101; Glu-Trp-Ile EWI motif-containing protein 101; Gm1016; Gm734; IGSF2; Immunoglobulin superfamily member 2; immunoglobulin superfamily, member 2; leukocyte surface protein; RP11-27K13.2; V7 | |
Rat | |
Protein A/G | |
RUO | |
630146 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction