Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD105, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137372
Description
Endoglin, also called CD105, is a homodimeric membrane glycoprotein primarily associated with human vascular endothelium. It is also found on bone marrow proerythroblasts, activated monocytes, and lymphoblasts in childhood leukemia. Endoglin is a component of the transforming growth factor beta receptor complex, as it binds TGFB1 with high affinity.
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.Specifications
CD105 | |
Polyclonal | |
Assay Dependent | |
P17813 | |
ENG | |
Recombinant protein encoding 26-176 aa of human CD105 | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
ENG | |
CD105, END, HHT1, ORW, ORW1, CD105 antigen, ENG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2022 | |
Store at 4°C, do not freeze | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction