Learn More
CD105 Mouse anti-Human, Brilliant Violet 650, Clone: 266, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563466
Catalog No. BDB563466
Add to cart
Description
Description
The 266 monoclonal antibody specifically binds to CD105. CD105 presents an integral membrane homodimer protein with subunits of 95 kDa found on vascular endothelial cells and syncytiotrophoblasts of placenta. CD105 is weakly expressed on stromal fibroblasts. It is also expressed on U937 cells, activated macrophages, and mesenchymal stem cells. CD105 is a component of the TGF-β receptor system in human umbilical vein endothelial cells and binds TGF-β1 and TGF-β3 with high affinity but does not bind toTGF-β2. Expression of CD105 is increased on activated endothelium in tissues undergoing angiogenesis, such as in tumors, or in cases of wound healing or dermal inflammation.
Specifications
Specifications
CD105
266
Endoglin; ENG; END; HHT1; ORW; ORW1
Affinity Purified
Human Umbilical Vein Endothelial Cells
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human