Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD106 Mouse anti-Human, PerCP-Cy5.5, Clone: 51-10C9, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563525

Catalog No. BDB563525


Add to cart

Description

Description

The 51-10C9 monoclonal antibody specifically binds to CD106. CD106 is a 100-110 kDa type I transmembrane sialoglycoprotrein that is also known as Vascular cell adhesion molecule-1 (VCAM-1) and INCAM-110. CD106 is expressed at high levels on the surface of cytokine-stimulated endothelium, and at minimal levels on unstimulated endothelium. VCAM-1 serves as a ligand for the leukocyte integrins α 4β 1 (CD49d/CD29 complex; VLA-4) and α 4β 7 (LPAM-1). The 51-10C9 monoclonal antibody inhibits the in vitro binding of lymphocytes and monocytes to VCAM-1 on stimulated endothelium.

Specifications

Specifications

CD106
51-10C9
VCAM-1; Vascular cell adhesion protein 1; INCAM-100; L1CAM
Affinity Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
PerCP-Cyanine5.5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit