Learn More
CD107b Rat anti-Mouse, Brilliant Violet 421, Clone: ABL-93, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 564249
Description
The ABL-93 monoclonal antibody specifically recognizes CD107b which is also known as Lysosome-associated membrane protein 2 (LAMP2/LAMP-2), Lysosomal membrane glycoprotein type B (LGP-B), and Mac-3. CD107b is a type I lysosomal transmembrane glycoprotein that serves as a useful marker to distinguish lysosomes from other cellular organelles. Mouse CD107b consists of a ∽40-kDa core protein which is heavily glycosylated to form heterogeneous mature glycoproteins of 110-120 kDa. It is principally expressed in epithelial cells and macrophages in a variety of organs in normal and Beige mutant mice.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
CD107b | |
Monoclonal | |
0.2mg/mL | |
Lamp2; LAMP-2; LAMP II; Lysosomal-associated membrane protein 2; LGP-B | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a λ |
Flow Cytometry, Immunocytochemistry | |
ABL-93 | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Glycoproteins purified from BALB/c mouse embryo 3T3 cell line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.