Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD107b Rat anti-Mouse, PerCP-Cy5.5, Clone: ABL-93, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564248

Catalog No. BDB564248


Add to cart

Description

Description

The ABL-93 monoclonal antibody specifically recognizes CD107b which is also known as Lysosome-associated membrane protein 2 (LAMP2/LAMP-2), Lysosomal membrane glycoprotein type B (LGP-B), and Mac-3. CD107b is a type I lysosomal transmembrane glycoprotein that serves as a useful marker to distinguish lysosomes from other cellular organelles. Mouse CD107b consists of a ~40-kDa core protein which is heavily glycosylated to form heterogeneous mature glycoproteins of 110-120 kDa. It is principally expressed in epithelial cells and macrophages in a variety of organs in normal and Beige mutant mice.

Specifications

Specifications

CD107b
ABL-93
PerCP-Cyanine5.5
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, λ
RUO
Primary
Murine
Flow Cytometry, Immunocytochemistry
0.2mg/mL
Lamp2; LAMP-2; LAMP II; Lysosomal-associated membrane protein 2; LGP-B
Affinity Purified
Glycoproteins purified from BALB/c mouse embryo 3T3 cell line
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit