Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD116 Mouse anti-Human, Brilliant Violet 650, Clone: hGMCSFR-M1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564044

Catalog No. BDB564044


Add to cart

Description

Description

The hGMCSFR-M1 antibody reacts with the subunit (GM-CSFR) of the human Granulocyte-Macrophage Colony-Stimulating Factor Receptor complex. This 75-85 kD subunit is also known as CD116. The hGMCSFR-M1 antibody was first clustered at the Fifth International Workshop on Human Leucocyte Differentiation Antigens. The GM-CSFR subunit associates with the 120-140 kD β c subunit (common subunit, CD131), that is shared with the receptors for interleukins IL-3 and IL-5. Both of the chains of the GM-CSFR complex are involved in ligand binding and intracellular signaling. The α chain appears to transmit most of the biological signals. CD116 is expressed by a variety of myeloid cell lines, hematopoietic and non-hematopoetic tumor cells, and normal cell types including monocytes, macrophages, neutrophils, eosinophils, myeloid dendritic cells, endothelial cells, fibroblasts, and placental trophoblasts. Lymphocytes are negative for GM-CSFR expression. Reports suggest that GM-CSFR plays a role in myeloid lineage growth and differentiation. The immunogen used to generate the hGMCSFR-M1 hybridoma was recombinant human GM-CSFR.

Specifications

Specifications

CD116
hGMCSFR-M1
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human
Flow Cytometry
0.2mg/mL
CSF2RA; GM-CSF Receptor alpha; GM-CSFRα; GMCSFRA; GMR, SMDP4
Affinity Purified
Recombinant human GM-CSFR
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit