Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD117/c-kit APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems FAB332A100
This item is not returnable.
View return policy
Description
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human samples. It is validated for Flow Cytometry.Specifications
CD117/c-kit | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, KIT, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCF R, SCFR, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
3815 | |
Human | |
Purified |
Flow Cytometry | |
47233 | |
Flow Cytometry 10 uL/10^6 cells | |
P10721 | |
KIT | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD117/c-kit Gln26-Thr520 Accession # P10721 | |
100 Tests | |
Primary | |
Detects human CD117/c-kit in ELISAs and Western blots. In sandwich immunoassays and Western blots, no cross-reactivity with recombinant mouse CD117 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction