Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117/c-kit Rat anti-Mouse, Clone: 180627, R&D Systems™

Rat Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen CD117/c-kit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD117/c-kit Gln25-Thr519 (Ala207Glu) Accession # P05532
Clone 180627
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1356SP
View Documents
R&D Systems
MAB1356SP
25 ug
Each for $125.00
Add to cart
 
MAB1356100
View Documents
R&D Systems
MAB1356100
100 ug
Each for $377.00
Add to cart
 
MAB1356500
View Documents
R&D Systems
MAB1356500
500 ug
Each for $839.00
Add to cart
 
Description

Description

CD117/c-kit Monoclonal antibody MAB1356 Specifically detects CD117/c-kit in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blotting
Specifications

Specifications

CD117/c-kit
180627
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Rat
IgG2a
3815
RUO
Mouse myeloma cell line NS0-derived recombinant mouse CD117/c-kit Gln25-Thr519 (Ala207Glu) Accession # P05532
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Purified
P05532
Monoclonal
Primary
Detects mouse CD117/c-kit in direct ELISAs.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit