Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Kit Mouse anti-Human, PE, Clone: NU-c-kit, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112194

Catalog No. 01-670-904

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD117 reacts with 8% of myelomonocytes and 75% of CD34-positive cells. The hematopoietic stem cells are included in this fraction and therefore, this gene product is considered to have an important function for differentiation and proliferation of hematopoietic cells. CD117 also reacts with placenta, brain, a part of acute myeloblastic leukemia cells and chronic myeloblastic leukemia cells, but not with acute lymphoblastic leukemia cells. MW is 145kDa.
Specifications

Specifications

c-Kit
Monoclonal
PE
KIT
Liquid
KIT
transfected cells with c-kit DNA in human leukemia cells (UT-7)
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
NU-c-kit
PBS with 0.1% sodium azide; pH 7.2
P10721
C-Kit, CD117, PBT, SCFR, mast/stem cell growth factor receptor Kit, p145 c-kit, piebald trait protein, proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, soluble KIT variant 1, tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein, ckit
Mouse
IgG1
100 Tests
4° C, do not freeze
Primary
3815
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit