Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a, Mouse anti-Human, Clone: HI111, BV421, BD

Manufacturer:  BD Biosciences 563936

Catalog No. BDB563936

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Host Species: Mouse
Clone: HI111
Isotype: IgG1, κ
Species Reactivity: Bb, Cyn, Hu, Rm

Flow cytometry

Specifications

Specifications

CD11a
Brilliant Violet 421
Murine
RUO
Monoclonal
HI111
Affinity Purified
100 Tests
Primary
Baboon, Cynomolgus, Human, Rhesus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit