Learn More
CD11A Mouse anti-Human, Brilliant Violet 510, Clone: HI111, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563479
 View more versions of this product
Catalog No. BDB563479
Add to cart
Description
Description
The HI111 monoclonal antibody specifically binds to CD11a, the 180 kDa integrin α chain. This type I transmembrane glycoprotein associates with CD18 (integrin β2) to form the heterodimeric glycoprotein CD11a/CD18. This heterodimer is also known as the lymphocyte (leukocytes) function associated antigen-1 (LFA-1) that is expressed on all leukocytes. LFA-1 is an adhesion molecule involved in lymphocyte and granulocyte functions. LFA-1 mediates adhesion of lymphoid cells to the vascular endothelium in association with its ligand, and the intracellular adhesion molecule-1 (ICAM-1), CD54. Other ligands are ICAM-2 (CD102) and ICAM-3 (CD50).
Specifications
Specifications
CD11a
HI111
LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL
Affinity Purified
IgG1, κ
RUO
Primary
Baboon, Cynomolgus, Human, Rhesus Monkey
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.