Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11A Mouse anti-Human, Brilliant Violet 510, Clone: HI111, BD

Mouse Monoclonal Antibody

$180.00 - $396.00

Specifications

Antigen CD11a
Isotype IgG1, κ
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Conjugate Brilliant Violet 510
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB563480
View Documents
BD Biosciences
563480
25 Tests
Each for $180.00
Add to cart
 
BDB563479
View Documents
BD Biosciences
563479
100 Tests
Each for $396.00
Add to cart
 
Description

Description

The HI111 monoclonal antibody specifically binds to CD11a, the 180 kDa integrin α chain. This type I transmembrane glycoprotein associates with CD18 (integrin β2) to form the heterodimeric glycoprotein CD11a/CD18. This heterodimer is also known as the lymphocyte (leukocytes) function associated antigen-1 (LFA-1) that is expressed on all leukocytes. LFA-1 is an adhesion molecule involved in lymphocyte and granulocyte functions. LFA-1 mediates adhesion of lymphoid cells to the vascular endothelium in association with its ligand, and the intracellular adhesion molecule-1 (ICAM-1), CD54. Other ligands are ICAM-2 (CD102) and ICAM-3 (CD50).

Specifications

Specifications

CD11a
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Mouse
Affinity Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
IgG1, κ
Brilliant Violet 510
Flow Cytometry
RUO
HI111
LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit