Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-CD11b/c, Clone: OX-42, BioLegend™ Non-distribution product as customer accommodation.

Manufacturer:  BiolegendSupplier Diversity Partner 201809

Catalog No. NC0193215



Description

Description

Host Species: Mouse
Clone: OX-42
Isotype: IgG2a, κ
Cross Reactivity: Rat

Flow Cytometry

Specifications

Specifications

CD11b/c
0.2mg/mL
Unpurified
IgG2a, κ
Primary
Rat
OX-42
APC
Murine
50μg
Monoclonal
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit