Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Hamster anti-Mouse, PE-CF594, Clone: HL3, BD

Hamster Monoclonal Antibody

Manufacturer:  BD Biosciences 562454

Catalog No. BDB562454


Add to cart

Description

Description

The HL3 monoclonal antibody specifically binds to the integrin α x chain of gp150, 95 (CD11c/CD18). CD11c is expressed on dendritic cells, CD4- CD8+ intestinal intraepithelial lymphocytes (IEL) and some NK cells. It is upregulated on IEL and lymph-node T cells following in vivo activation. Cells of the monocyte/macrophage lineage have been reported to express low levels of CD11c. CD11c plays a role in binding of iC3b.

Specifications

Specifications

CD11c
HL3
PE-CF594
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Hamster
IgG1, λ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4
Affinity Purified
C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
0.1mg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit