Learn More
anti-CD11c, PE, Clone: N418, R&D Systems™
Manufacturer:  R&D Systems FAB69501P100
 View more versions of this product
Catalog No. FAB69501P10
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD11c
N418
Supplied in a saline solution containing BSA and Sodium Azide.
Hamster
IgG
100 Tests
Primary
16411
Murine
Flow Cytometry
PE
Purified
Mouse spleen dendritic cells
Protein A or G purified from hybridoma culture supernatant
RUO
Monoclonal
Detects mouse CD11c.