Learn More
CD11c Mouse anti-Human, PerCP-Cy5.5, Clone: B-ly6, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565227
Catalog No. BDB565227
Add to cart
Description
Description
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.
Specifications
Specifications
CD11c
B-ly6
ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6
Affinity purified
Spleen Cells from Human with Hairy Cell Leukemia
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PerCP-Cyanine5.5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
IgG1, κ
RUO
Primary
Human