Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1R2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA135856

Catalog No. 01-672-357


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

The IL-1 system includes two agonists (IL-1· and IL-1‚), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
Specifications

Specifications

IL1R2
Polyclonal
Unconjugated
IL1R2
Liquid
IL1R2
Recombit human IL.
Purified
RUO
Antibody
Polyclonal
Human
ELISA
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P27930
CD121b, IL1RB, CD121 antigen-like family member B, CDw121b, IL-1 type II receptor, IL-1R-2, IL-1R-beta, IL-1RT-2, IL-1RT2, antigen CDw121b, interleukin-1 receptor beta, interleukin-1 receptor type 2, interleukin-1 receptor type II, type II interleukin-1 receptor, beta, ILR2
Rabbit
IgG
100 μg
4° C
Primary
7850
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit