Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD127 (IL-7Ra) Violer Fluor 450, Clone: A7R34,

Rat Monoclonal Antibody

$153.50 - $461.00

Specifications

Antigen CD127 (IL-7Ra)
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1511MI
View Documents
MilliporeSigma™
MABF1511
25μg
Each for $153.50
Add to cart
 
MABF1512MI
View Documents
MilliporeSigma™
MABF1512
100μg
Each for $461.00
Add to cart
 
Description

Description

Specifically detects CD127 (IL-7Ra) Clone: A7R34 in Mouse samples, and it is validated for Flow Cytometry

The A7R34 antibody is specific for mouse CD127, a 60-90kDa cell surface protein also known as the Interleukin-7 Receptor alpha chain, or IL-7R alpha. CD127 is typically expressed at the cell surface as a heterodimer with the common gamma chain (CD132). This complex acts as the functional receptor for IL-7, a cytokine important in T and B cell development, and in mature T cell homeostasis. A second cytokine known as Thymic Stromal Lymphopoietin (TSLP) also binds to a receptor complex of CD127 and the TSLPR chain to trigger activation of dendritic cells, and is involved in B cell development, allergy and autoimmunity.The A7R34 antibody may be used as a phenotypic marker for CD127 on immature B cells, on subsets of thymocytes which are double negative (CD4-CD8- ) or single positive (CD4+ or CD8+), and at low levels on mature, peripheral T cells. CD127 is a key marker, when used in combination with CD4 and CD25, to distinguish Treg and effector/memory Treg populations known as T(REM).
Specifications

Specifications

CD127 (IL-7Ra)
Monoclonal
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
Proprietary immunogen
Il7r
RUO
Purified by size-exclusion chromatography
Primary
Purified
A7R34
Rat
P16872
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit