Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD13 Rat anti-Mouse, Alexa Fluor 647, Clone: R3-242, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564352

Catalog No. BDB564352


Add to cart

Description

Description

The R3-242 monoclonal antibody specifically binds to aminopeptidase N (APN), which has been reported to be the mouse CD13 homologue. APN is expressed in a variety of mammalian tissues, including intestinal and renal epithelial brush border, endothelium, liver, lung, and brain. In lymphoid tissues, R3-242 mAb is reported to detect APN on monocytes, macrophages, mast cells, dendritic cells, and B lymphocytes, but not on T cells, and thymocytes. However, the reported reactivity with B cells may be non-specific, via binding to the Fc γreceptors (CD16/CD32). This observation is in agreement with reports using different antibodies to mouse APN . APN may play a role in antigen presentation in the immune system. R3-242 mAb does not inhibit the enzymatic activity of APN.

Specifications

Specifications

CD13
R3-242
Alexa Fluor 647
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG1
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Anpep; Alanyl aminopeptidase; Aminopeptidase N; AP-N; mAPN; Lap-1
Affinity Purified
Crude Aminopeptidase N Preparation from BALB/c Mouse Intestinal Epithelium
0.1mg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit