Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD13, Clone: 366 (MY7), Beckman Coulter™

$746.64

Specifications

Antigen CD13
Format Purified
Host Species Murine
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CO6602989
View Documents
Coulter Corporation
6602989
100 Tests
Each for $746.64
Add to cart
 
Description

Description

The CD13 antigen is a transmembrane glycoprotein with a large extracellular region and a small intracellular NH2-terminal tail. It has a molecular weight of 150kDa and is expressed on the cell surface as a non-covalently linked homodimer. It is found on most cells of myeloid origin including neutrophils, eosinophils, basophils and monocytes from normal peripheral blood. It is absent from B and T lymphocytes as well as from red blood cells and platelets. This molecule is found on the surface of committed progenitor cells defined as the Granulocyte-Monocyte Colony Forming Units (CFU-GM) in normal bone marrow. There are up to five subpopulations of CD13 molecules possessing different levels of glycosylation, which may explain the different binding patterns of various CD13 antibodies.

Host Species: Mouse
Clone: 366 (MY7)
Isotype: IgG1
Species Reactivity: Human

Flow Cytometry, Immunohistochemistry, Immunoprecipitation

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

CD13
Murine
Monoclonal
Purified
IgG1
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit