Learn More
CD13 Mouse anti-Human, Clone: E735, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112033
Catalog No. 01-670-756
Add to cart
Description
Description
CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.
Specifications
Specifications
CD13
Monoclonal
Unconjugated
ANPEP
Liquid
ANPEP
human cell line KG-1a
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
E735
PBS with 0.08% sodium azide
P15144
AP-M, AP-N, Apn, Cd13, P150, alanyl aminopeptidase, aminopeptidase M, aminopeptidase N, aminopeptidase N/CD13, membrane protein p161, microsomal aminopeptidase
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
290