Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP130 Mouse anti-Human, Clone: 28126, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523817

Catalog No. PIMA523817


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human (rh) IL-2 R, rhIL-4 R, or rhIL-6 R is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications

Specifications

GP130
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P40189
CD130, CDw130, GP130, GP130-RAPS, IL6R-beta, IL6ST
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3572
Flow Cytometry, Neutralization
28126
Unconjugated
IL6ST
Lyophilized
IL6ST
Sf 21-derived recombit human gp130 extracellular domain
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit