Learn More
CD131 Mouse anti-Human, Alexa Fluor 647, Clone: 3D7, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564191
Description
The 3D7 antibody reacts with CD131, the 120 kD common β chain (βc) which is shared with the receptor complexes for human granulocyte-macrophage colony stimulating factor (GM-CSFR), interleukin-3 (IL-3R) and interleukin-5 (IL-5R). Together with the α subunit of either the IL-3R (IL-3Rα), IL-5R (IL-5Rα), or GM- CSFR (GM-CSFRα), the common ββ chains.

Specifications
CD131 | |
Monoclonal | |
Alexa Fluor 647 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
3D7 | |
Cytokine receptor common β chain; CDw131; IL-3Rβ; IL-5Rβ | |
Human CD131 Transfected COS Cells | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.