Learn More
CD131 Mouse anti-Human, Brilliant Violet 421, Clone: 3D7, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564192
Catalog No. BDB564192
Add to cart
Description
Description
The 3D7 antibody reacts with CD131, the 120 kD common β chain ( β c) which is shared with the receptor complexes for human granulocyte-macrophage colony stimulating factor (GM-CSFR), interleukin-3 (IL-3R) and interleukin-5 (IL-5R). Together with the α subunit of either the IL-3R (IL-3R α ), IL-5R (IL-5R α ), or GM- CSFR (GM-CSFR α ), the common β β chains.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Specifications
Specifications
CD131
3D7
Cytokine receptor common β chain; CDw131; IL-3Rβ; IL-5Rβ
Affinity Purified
Human CD131 Transfected COS Cells
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human