Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD132 Rat anti-Human, Brilliant Violet 650, Clone: TUGH4, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 563406

Catalog No. BDB563406


Add to cart

Description

Description

The TUGh4 monoclonal antibody specifically binds to CD132. CD132 is a 65-70 kDa type 1 transmembrane glycoprotein that is encoded by the IL2RG (interleukin 2 receptor, gamma) gene. CD132 is also known as the common γsubunit (γc) and is shared by the IL-2, IL-4, IL-7, IL-9, IL-15 and IL-21 receptor complexes. CD132 is broadly expressed by most peripheral T and B lymphocytes, NK cells, monocytes, and granulocytes. The cytoplasmic domain of the γc chain plays an important role in cytokine-mediated signal transduction. Mutation of the IL2RG gene results in X-linked severe combined immunodeficiency (XSCID). The TUGh4 antibody recognizes a different epitope from that recognized by the CD132-specific clone, AG184.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will bespillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD132
TUGH4
IL-2RG; IL-2Rγ; γc; Common γ chain; Common gamma chain; gamma c; SCIDX1
Affinity Purified
Human γc Transfected Cell Line
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2b, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit