Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40 Monoclonal Antibody (443318), PE, Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA523676
Description
In Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFAS, rhGITR, rhHVEM, rhLT beta R, rhNGF R, rhRANK, rhRELT, rhTAJ, rhTNF RI, rhTNF RII, rhTRAIL, rhTRAIL R4, rhTRAIL R3, rhTWEAK R, or rhXEDAR is observed.
OX40 Monoclonal antibody specifically detects OX40 in Human samples. It is validated for Flow Cytometry.Specifications
OX40 | |
Monoclonal | |
PE | |
Tnfrsf4 | |
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
Rat | |
Protein A/G | |
RUO | |
7293 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
443318 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P43489 | |
Tnfrsf4 | |
Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4 Leu29-Ala216 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction