Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD138 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137379

Catalog No. 01-673-251



Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil tissue.

CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is lost from the apoptotic myeloma cells.
Specifications

Specifications

CD138
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P18827
SDC1, SYND1, SDC, CD antigen 138
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6382
Immunohistochemistry (Paraffin)
Unconjugated
SDC1
Liquid
SDC1
Recombit protein encoding aa 18-218 human CD138.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit