Learn More
CD14 Mouse anti-Human, Alexa Fluor 700, Clone: M5E2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 561029
Description
Reacts with the 53-55kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
Host Species: Mouse
Clone: M5E2
Isotype: IgG2a, κ
Cross Reactivity: Human, Rhesus, Cynomolgus, Baboon
Flow Cytometry
Specifications
CD14 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Human CD14 Protein | |
25 μg | |
Immunology | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
IgG2a κ |
Flow Cytometry | |
M5E2 | |
Alexa Fluor 700 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.