Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14, APC, clone: FWKW, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112257

Catalog No. 01-670-953

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD14 is a single-copy gene encoding 2 protein forms: a 50- to 55-kD glycosylphosphatidylinositol-anchored membrane protein (mCD14) and a monocyte or liver-derived soluble serum protein (sCD14) that lacks the anchor. Both molecules are critical for lipopolysaccharide (LPS)-dependent signal transduction, and sCD14 confers LPS sensitivity to cells lacking mCD14. Increased sCD14 levels are associated with inflammatory infectious diseases and high mortality in gram-negative shock.

Specifications

Specifications

CD14
Monoclonal
APC
CD14
Purified
CD14
White blood cells.
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
FWKW
PBS with 0.2% carrier protein and 0.08% sodium azide
P08571
monocyte differentiation antigen CD14, myeloid cell-specific leucine-rich glycoprotein
Murine
IgG2b
100 Tests
Store at 4°C
Primary
929
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit