Learn More
CD13/CD14 Mouse anti-Human, FITC, PE, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112204
Catalog No. 01-670-914
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD14 (clone FWKW, isotype IgG2b) and PE-conjugated CD13 (clone E735, isotype IgG1).
CD14 is a single-copy gene encoding 2 protein forms: a 50- to 55-kD glycosylphosphatidylinositol-anchored membrane protein (mCD14) and a monocyte or liver-derived soluble serum protein (sCD14) that lacks the anchor. Both molecules are critical for lipopolysaccharide (LPS)-dependent signal transduction, and sCD14 confers LPS sensitivity to cells lacking mCD14. Increased sCD14 levels are associated with inflammatory infectious diseases and high mortality in gram-negative shock. This antibody recognizes the human CD13 cell surface glycoprotein, a 150kD molecule expressed by granulocytes and monocytes, and by myeloid leukemia cells. Inhibits infection of cells by human coronavirus, inhibits aminopeptidase N activity of the CD13 molecule immunoprecipitates. MA1-22542 detects CD13 in Human samples.
Specifications
Specifications
CD13/CD14
Cocktail
FITC, PE
ANPEP
Liquid
ANPEP, CD14
human cell line KG-1a
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
FWKW, E735
PBS with 0.2% carrier protein and 0.08% sodium azide
P08571, P15144
ANPEP, AP-M, AP-N, Apn, Cd13, P150, alanyl aminopeptidase, aminopeptidase M, aminopeptidase N, aminopeptidase N/CD13, membrane protein p161, microsomal aminopeptidase, monocyte differentiation antigen CD14, myeloid cell-specific leucine-rich glycoprotein
Mouse
IgG1/IgG2b
50 Tests
4° C, do not freeze
Primary
290, 929